accessibility menu, dialog, popup

UserName

OOOSmSm

CAS RN 12060-58-1

IUPAC Name: disamarium(3+) trioxidandiide
Synonyms: samarium(III) oxide
Molecular Weight (g/mol): 348.72
Molecular Formula: O3Sm2
InChi Key: FKTOIHSPIPYAPE-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[Sm+3].[Sm+3]
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)