missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium ICP Standard, 1000 ppm Ru in 10% HCl, Ricca Chemical

Catalog No. PRU1KH100
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
PRU1KH100 100 mL Natural LDPE Bottle
PRU1KH500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRU1KH100 Supplier Ricca Chemical Company Supplier No. PRU1KH100

Chemical Identifiers

CAS 14898-67-0 , 7647-01-0 , 7732-18-5
Molecular Formula Cl3Ru
Molecular Weight (g/mol) 207.42
MDL Number MFCD00149844
InChI Key YBCAZPLXEGKKFM-UHFFFAOYSA-K
IUPAC Name ruthenium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Ru+3]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Ru
CAS Min % 87.0, 10.32, 0.21
CAS Max % 91.0, 10.74, 0.22
Equivalence 1 mL = 1 mg Ru
Physical Form Liquid
Color Brown/Red
Melting Point Approximately 0°C
Type Standard
Quantity 100 mL
Chemical Name or Material Ruthenium ICP Standard
Show More Show Less