missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Ringer's Solution, lactate-buffered
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
Spécifications
| Chemical Name or Material | Ringer's Solution |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Synonym | dihydrogen oxide, dihydrogen monoxide |
| Packaging | Plastic bottle |
RUO – Research Use Only