Learn More
Rhodium(III) chloride hydrate, Premion™, 99.99% (metals basis), Thermo Scientific™
$9,671.34
Identifiants chimiques
| CAS | 20765-98-4 |
|---|---|
| Molecular Formula | RhCl3·xH2O |
| MDL Number | MFCD00149839 |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
|
AA4370206
|
N/A |
chaque for $9,671.34
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identifiants chimiques
| 20765-98-4 | |
| RhCl3·xH2O | |
| MFCD00149839 |
Spécifications
| 20765-98-4 | |
| MFCD00149839 | |
| Soluble in water and alcohol. Slightly soluble in acetone. | |
| 99.99% | |
| Hygroscopic | |
| Rhodium (III) chloride hydrate |
| RhCl3·xH2O | |
| 14,8188 | |
| 209.26 (Anhydrous) | |
| Premion™ | |
| (metals basis) |
Sécurité et manipulation
GHS H Statement
H318-H290-H341-H302
Causes serious eye damage.
May be corrosive to metals.
Suspected of causing genetic defects.
Harmful if swallowed.
P201-P202-P234-P264b-P270-P281-P301+P312-P305+P351+P338-P308+P313-P310-P330-P390-P501c
H290-H302-H318-H341
DOTInformation : Hazard Class: 8; Packaging Group: III
EINECSNumber : 233-165-4
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only