missing translation for 'onlineSavingsMsg'
Learn More

Rhodium(III) chloride hydrate, Premion™, 99.99% (metals basis), Thermo Scientific™

Catalog No. AA4370206
Change view
Click to view available options
Quantity:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4370206 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4370206 Supplier Thermo Scientific Chemicals Supplier No. 04370206

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 20765-98-4
Molecular Formula RhCl3·xH2O
MDL Number MFCD00149839

Specifications

Quantity 5g
Merck Index 14,8188
Solubility Information Soluble in water and alcohol. Slightly soluble in acetone.
Formula Weight 209.26 (Anhydrous)
Percent Purity 99.99%
Grade Premion™
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Rhodium (III) chloride hydrate
Hazard Category H290-H302-H318-H341
Hazard Statement GHS H Statement
H318-H290-H341-H302
Causes serious eye damage.
May be corrosive to metals.
Suspected of causing genetic defects.
Harmful if swallowed.
Precautionary Statement P201-P202-P234-P264b-P270-P281-P301+P312-P305+P351+P338-P308+P313-P310-P330-P390-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 233-165-4
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only