Learn More
Corning™ PYREX™ Reusable Heavy-Wall Ignition Tubes

For high-temperature work in the classroom
$1,898.87
Spécifications
| Design Type | Plain |
|---|---|
| Material | Borosilicate Glass |
| For Use With | For high-temperature work in the classroom |
| Sterility | Non-sterile |
| Graduated | No |
| Numéro de catalogue | Numéro du manufacturier. | Diameter (Metric) | Length (Metric) | Prix | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Diameter (Metric) | Length (Metric) | Prix | Quantity & Availability | ||||
14960F
![]() |
Corning™
986025 |
25 mm | 200 mm |
caisse de 120 chaque for $1,898.87
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
Description
These new culture tubes with flat bottoms are ideal for plant tissue culture and general culture work. The flat bottom facilitates use with agar slants. Walls and bottoms are of uniform thickness. The tubes are rimless for greater convenience in plugging and rack storage. Optional polypropylene two position closure. Position closure open for gas exchange or closed for humidified environment. Internal drip ring minimizes contamination.
For closures, see Cat. Nos 26796-38, 26797-38, 26794-25, 26795-25, 26798-40, and 26799-40.
Spécifications
| Plain | |
| For high-temperature work in the classroom | |
| No |
| Borosilicate Glass | |
| Non-sterile | |
| Culture Tube |