missing translation for 'onlineSavingsMsg'
Learn More

Corning™ PYREX™ Reusable Heavy-Wall Ignition Tubes
GREENER_CHOICE

Catalog No. 14960F
Change view
Click to view available options
Diameter (Metric):
25 mm
Length (Metric):
200 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Length (Metric)
14960F 25 mm 200 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14960F Supplier Corning™ Supplier No. 986025

For high-temperature work in the classroom

These new culture tubes with flat bottoms are ideal for plant tissue culture and general culture work. The flat bottom facilitates use with agar slants. Walls and bottoms are of uniform thickness. The tubes are rimless for greater convenience in plugging and rack storage. Optional polypropylene two position closure. Position closure open for gas exchange or closed for humidified environment. Internal drip ring minimizes contamination.

For closures, see Cat. Nos 26796-38, 26797-38, 26794-25, 26795-25, 26798-40, and 26799-40.

 

Specifications

Design Type Plain
Length (Metric) 200 mm
Material Borosilicate Glass
For Use With For high-temperature work in the classroom
Sterility Non-sterile
Graduated No
Diameter (Metric) 25 mm
Product Type Culture Tube