missing translation for 'onlineSavingsMsg'
Learn More

CritiCore Protective Wear Reusable Bouffant Cap in White Taffeta

Catalog No. 17012831
Change view
Click to view available options
Size:
18 in.
19.5 in.
22 in.
24 in.
26 in.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17012831 26 in.
17012827 18 in.
17012828 19.5 in.
17012829 22 in.
17012830 24 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17012831 Supplier CritiCore Protective Wear Supplier No. 11BF0581TAFF012XLWHT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This reusable bouffant cap in white taffeta is ideal for any cleanrooms that do not require ESD. Lightweight and durable with a tunneled elastic opening.

  • Tunneled elastic opening
  • Taffeta fabric
  • Primary applications: cleanroom pharmaceutical, biotechnological, medical device manufacturing

Specifications

Product Type Bouffant
Size 26 in.
Material Polyester
Color White
Disposable Reusable