missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Tubing Reamer

Catalog No. 06710510
Encompass
Change view
Click to view available options
For Use With (Equipment):
6.4 or 3.2 mm Tubing
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
06710510 6.4 or 3.2 mm Tubing
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06710510 Supplier Restek™ Supplier No. RK 20134

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Removes burrs from stainless steel tubing

  • For 6.4 or 3.2mm (1/4 or 1/8 in.) tubing
  • Nonslip safety design

Specifications

Type Tubing Reamer
For Use With (Equipment) 6.4 or 3.2 mm Tubing