missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Restek™ Sulfinert Gas Sampling Valves
Sulfinert-treated valves are ideal for samples containing low concentrations of sulfur or other active compounds.
Specifications
| For Use With (Equipment) | 1/16 in. Fittings |
|---|---|
| Material | Sulfinert-coated |
| Type | Manual Sample Valve |
Description
- “W Type” Valve
- 0.0625 in. fittings
- 0.40mm Port diameter
Specifications
| 1/16 in. Fittings | |
| Sulfinert-coated | |
| Manual Sample Valve |