Learn More
Restek™ Sulfinert Gas Sampling Valves
Sulfinert-treated valves are ideal for samples containing low concentrations of sulfur or other active compounds.
Supplier:  Restek™ RK 20585
Encompass
 View more versions of this product
Catalog No. 06710697
Only null left
Description
Description
  • “W Type” Valve
  • 0.0625 in. fittings
  • 0.40mm Port diameter
Specifications
Chemical Identifiers
1/16 in. Fittings
Sulfinert-coated
Manual Sample Valve
Specifications
1/16 in. Fittings
Sulfinert-coated
6-Port
Manual Sample Valve
Product Suggestions
Product Suggestions