Learn More
Restek™ Rtx™-1301 Capillary Columns, G43 Equivalent
For residual solvents, alcohols, oxygenates, and volatile organic compounds
$1697.75 - $2043.81
| Catalog Number | Mfr. No. | Length (Metric) | Diameter (Metric) Inner | Film Thickness | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Length (Metric) | Diameter (Metric) Inner | Film Thickness | Price | Quantity | |||
06710222
|
Restek™
RK16008 |
30 m | 0.25 mm | 0.1 μm |
Each for $1,697.75
|
|
|||
06710251
|
Restek™
RK16040 |
30 m | 0.53 mm | 0.5 μm |
Each for $2,043.81
|
|
|||
06710263
|
Restek™
RK16053 |
30 m | 0.25 mm | 1 μm |
Each for $1,711.70
|
|
|||
06710268
|
Restek™
RK16055 |
30 m | 0.53 mm | 1 μm |
Each for $1,932.21
|
|
|||
06710279
|
Restek™
RK16069 |
30 m | 0.32 mm | 1.5 μm |
Each for $1,721.42
|
|
|||
06710282
|
Restek™
RK16070 |
30 m | 0.53 mm | 1.5 μm |
Each for $2,043.81
|
|
|||
06710290
|
Restek™
RK16085 |
30 m | 0.53 mm | 3 μm |
Each for $1,932.21
|
|
|||
Description
- One of the best cyanosiloxane bonded stationary phases available
- No other column manufacturer provides lower bleed, longer lifetimes, or better inertness
- Polymer is fully characterized to ensure long-term reproducibility, column-to-column consistency, and low bleed
Order Info
Custom lengths and film thicknesses available.
Specifications
| G43 | |
| 350°C | |
| Fused Silica |