Learn More
Restek™ Rtx™-1301 Capillary Columns, G43 Equivalent DFS Item
For residual solvents, alcohols, oxygenates, and volatile organic compounds
Supplier:  Restek™ RK16085
 View more versions of this product
Catalog No. 06710290
Only null left
Explore available promotions
Description
Description
  • One of the best cyanosiloxane bonded stationary phases available
  • No other column manufacturer provides lower bleed, longer lifetimes, or better inertness
  • Polymer is fully characterized to ensure long-term reproducibility, column-to-column consistency, and low bleed
Order Info
Custom lengths and film thicknesses available.
Specifications
Specifications
0.53 mm
Rtx™-1301 (G43)
Crossbond™, Oxygenates Analysis, Residual Solvents Analysis, Volatile Organic Compounds Analysis
-20°C to +240°C
30 m
Low to mid polar
G43
350°C
3 μm
3 μm
Stainless Steel
Fused Silica
Product Suggestions
Product Suggestions