missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Replacement Valve for SGE Gas-Tight Syringes Fitted with Removable Needle Valve

Catalog No. 06712555
Encompass
Change view
Click to view available options
Volume (Metric):
25 μL to 2.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
06712555 25 μL to 2.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06712555 Supplier Restek™ Supplier No. RK 24898

For gas-tight syringes with shut-off valves.

  • Model V syringe valve
  • Fits any SGE removable needle syringe with volume range 25μL to 2.5mL
  • Gauge: 23

Specifications

Needle Gauge 23 G
Length (English) Needle 2 in.
Product Type Syringe Valve
Quantity Ea.
Volume (Metric) 25 μL to 2.5 mL
Needle Point Style Cone
For Use With (Equipment) SGE Removable Needle Syringes
Connection Type Removable