missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Research Grade Fetal Bovine Serum, Canadian Sourced
Description
- Canadian origin
- Compliant with current good manufacturing practices (cGMP)
- For research use only
Spécifications
Spécifications
| Additional Assays | Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, Glutamic Pyruvic Transaminase (SGPT), Glutamic Oxaloacetic Transaminase (SGOT), pH, Total Protein (Ph. Eur. 2.5.33), Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality (Ph. Eur. 2.2.35), Iron, Total Iron Binding Capacity (TIBC), Percent Saturation, Glucose, IgG – Nephelometer |
| Content And Storage | ≤ -10°C |
| Country of Origin | Canada |
| Form | Liquid |
| Quantity | 500 mL |
| Grade | Research |
| Processing | Yes |
| Heat Inactivated | No |
| Hemoglobin Concentration | < 35 mg/dL |
| Lyophilized | No |