Learn More
Contec Replacement Collars
Interchangeable mop handles, available in carbon fiber, anodised aluminum, stainless steel, or fiberglass.
$13.05 - $62.93
Spécifications
Product Type Cleanroom Mop Handle
Sterility Non-sterile
Autoclavable Autoclavable
Color-coded No
Material Carbon Fiber
voir les spécifications
Produits 3
Numéro de catalogue Numéro du manufacturier. Includes Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Includes Prix Quantity & Availability  
17720592
Afficher les documents Détails de la promotion
Contec
L3270114
1 Collar Pole
chaque for $13.05
Il en reste null
17720591
Afficher les documents Détails de la promotion
Contec
L224
3 Collar Pole
chaque for $37.76
Il en reste null
17720593
Afficher les documents Détails de la promotion
Contec
L334
5 Collar Pole
chaque for $62.93
Il en reste null
Description
Description
  • Compatible with all QuickConnect™ frames
  • Multiple lengths available
  • Fully autoclavable QuickConnect™ mop handles and interchangeable mop head frames work together for maximum flexibility when cleaning in critical environments.
  • Allows investment in one handle with various interchangeable heads required for specific applications.
  • Handles are available in several lengths and styles.
  • Telescoping handles are available in lightweight carbon fiber, anodised aluminum, durable stainless steel, or fiberglass.
  • Choose from a variety of lengths to suit different cleaning needs.
Spécifications
Spécifications
Cleanroom Mop Handle
Autoclavable
Carbon Fiber
Non-sterile
No
Videos
FDS