Learn More
Contec Replacement Collars
Interchangeable mop handles, available in carbon fiber, anodised aluminum, stainless steel, or fiberglass.
Supplier: Contec L334
Description
- Compatible with all QuickConnect™ frames
- Multiple lengths available
- Fully autoclavable QuickConnect™ mop handles and interchangeable mop head frames work together for maximum flexibility when cleaning in critical environments.
- Allows investment in one handle with various interchangeable heads required for specific applications.
- Handles are available in several lengths and styles.
- Telescoping handles are available in lightweight carbon fiber, anodised aluminum, durable stainless steel, or fiberglass.
- Choose from a variety of lengths to suit different cleaning needs.
Identifiants chimiques
| Cleanroom Mop Handle | |
| Autoclavable | |
| Carbon Fiber |
| Non-sterile | |
| No |
Spécifications
| Cleanroom Mop Handle | |
| Autoclavable | |
| No |
| Non-sterile | |
| 5 Collar Pole | |
| Carbon Fiber |