Learn More
Fisherbrand™ Recessed Safety Station
Description
- Recessed barrier-free eye/face wash and shower safety station with ceiling or wall mounted exposed shower head and drain pan.
- 10 in. diameter stainless steel shower head with 20 GPM flow control.
- 1 in. IPS brass stay-open shower ball valve with stainless steel 'panic bar' actuator.
Spécifications
Spécifications
| Product Type | Safety Station |
| Material | Brass |
| Mounting Type | Ceiling |