missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ReadyProbes™ Solvent-Resistant Permanent Marking Pen

Catalog No. R37623
Change view
Click to view available options
Quantity:
5 pens
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R37623 5 pens
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R37623 Supplier Invitrogen™ Supplier No. R37623

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Enables solvent-resistant writing on frosted glass microscope slides.

  • The ReadyProbes Solvent-Resistant Permanent Marking Pen enables solvent-resistant writing on frosted glass microscope slides.
  • It is resistant to most solvents, including alcohols and xylene.
Note: This marking pen enables solvent-resistant writing on the frosted end of glass microscope slides. Plain glass (unfrosted) or smooth plastic are not considered appropriate surfaces to retain the pen ink as the residue can be easily wiped off of these surfaces. Using the pen on other types of hard surfaces (such as smooth cryo-vial surface or Eppendorf tubes) could produce writing that is not solvent-resistent. When used on frosted glass microscope slides, ReadyProbes Solvent-Resistant Permanent Marking Pen is resistant to most solvents, including alcohols and xylene. Be sure to let the ink dry before exposing to solvent.

Specifications

Type Permanent Marking Pen
Product Line ReadyProbes
Content And Storage A set of 5 pens.

Store at room temperature.

When stored as directed, product can be used for 6 months after receipt.

Includes 5 Pens
Quantity 5 pens
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
When would I use the ReadyProbes Solvent-Resistant Permanent Marking Pen versus a standard hydrophobic barrier pen?

We recommend using the ReadyProbes Solvent-Resistant Permanent Marking Pen if you will be subjecting your samples to any solutions containing alcohol or xylene.

For Research Use Only. Not for use in diagnostic procedures.