missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spécifications
| Accession Number | C230052L06Rik; DFNB39; fibroblast-derived tumor cytotoxic factor; F-TCF; hepatocyte growth factor; hepatocyte growth factor (hepapoietin A; scatter factor); Hepatocyte growth factor alpha chain; Hepatocyte growth factor beta chain; Hepatopoeitin A; hepatopoeitin-A; HepatopoietinA; hepatopoietin-A; Hgf; HGF/SF; HGFB; HPTA; lung fibroblast-derived mitogen; NK1; NK2; Scatter factor; SF; SF/HGF |
|---|---|
| Assay Range | 0.8 to 200 ng/mL |
| Assay Sensitivity | 0.8 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
ERA21RBX5
|
Invitrogen™
ERA21RBX5 |
5 x 96 Tests |
chaque for $4,420.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
ERA21RB
|
Invitrogen™
ERA21RB |
96 Tests |
chaque for $938.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
Rat HGF quantitates rat HGF in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant rat HGF.
Hepatocyte growth factor regulates cell growth, cell motility, and morphogenesis by activating a tyrosine kinase signaling cascade after binding to the proto-oncogenic c-Met receptor. Hepatocyte growth factor is secreted by mesenchymal cells and acts as a multi-functional cytokine on cells of mainly epithelial origin. Its ability to stimulate mitogenesis, cell motility, and matrix invasion gives it a central role in angiogenesis, tumorogenesis, and tissue regeneration. It is secreted as a single inactive polypeptide and is cleaved by serine proteases into a 69-kDa alpha-chain and 34-kDa beta-chain. A disulfide bond between the alpha and beta chains produces the active, heterodimeric molecule. The protein belongs to the plasminogen subfamily of S1 peptidases but has no detectable protease activity. Alternative splicing of this gene produces multiple transcript variants encoding different isoforms.Spécifications
| C230052L06Rik; DFNB39; fibroblast-derived tumor cytotoxic factor; F-TCF; hepatocyte growth factor; hepatocyte growth factor (hepapoietin A; scatter factor); Hepatocyte growth factor alpha chain; Hepatocyte growth factor beta chain; Hepatopoeitin A; hepatopoeitin-A; HepatopoietinA; hepatopoietin-A; Hgf; HGF/SF; HGFB; HPTA; lung fibroblast-derived mitogen; NK1; NK2; Scatter factor; SF; SF/HGF | |
| 0.8 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| Hgf | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| HGF | |
| 4 hrs 45 mins |
| 0.8 to 200 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 24446 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |