missing translation for 'onlineSavingsMsg'
Learn More

Rapamycin 98.0+%, TCI America™

Catalog No. R009725MG
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R009725MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R009725MG Supplier TCI America Supplier No. R009725MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 53123-88-9
Molecular Formula C51H79NO13
Molecular Weight (g/mol) 914.187
MDL Number MFCD00867594
InChI Key QFJCIRLUMZQUOT-HPLJOQBZSA-N
PubChem CID 5284616
ChEBI CHEBI:9168
SMILES CC1CCC2CC(C(=CC=CC=CC(CC(C(=O)C(C(C(=CC(C(=O)CC(OC(=O)C3CCCCN3C(=O)C(=O)C1(O2)O)C(C)CC4CCC(C(C4)OC)O)C)C)O)OC)C)C)C)OC

Specifications

Type Rapamycin
Physical Form Crystalline Powder
Melting Point 177°C
Color White/Yellow
Quantity 25 mg
Formula Weight 914.19
Percent Purity ≥98.0% (HPLC)
Chemical Name or Material Rapamycin
RTECSNumber VE6250000
TSCA No
Recommended Storage Freezer