missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Raloxifene hydrochloride

CAS: 82640-04-8 | C28H28ClNO4S, C28H27NO4S·HCl | 510.05 g/mol

$463.91 - $1245.11

Chemical Identifiers

CAS 82640-04-8
Molecular Formula C28H27NO4S·HCl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC466500010
View Documents
Thermo Scientific Chemicals
466500010
1 g
Each for $463.91
Only null left
 
AC466500050
View Documents
Thermo Scientific Chemicals
466500050
5 g
Each for $1,245.11
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

82640-04-8
C28H27NO4S·HCl

Specifications

82640-04-8
Solubility aq. soln.: .. Other solubilities: soluble in dmso
C28H27NO4S·HCl
BKXVVCILCIUCLG-UHFFFAOYSA-N
hydrogen 2-(4-hydroxyphenyl)-3-{4-[2-(piperidin-1-yl)ethoxy]benzoyl}-1-benzothiophen-6-ol chloride
Raloxifene hydrochloride
1 g
263.0°C
510.04
[H+].[Cl-].OC1=CC=C(C=C1)C1=C(C(=O)C2=CC=C(OCCN3CCCCC3)C=C2)C2=CC=C(O)C=C2S1
510.05
Videos
Safety and Handling

Safety and Handling

GHS H Statement Suspected of causing cancer.

May damage fertility or the unborn child.

GHS P Statement Obtain special instructions before use.

Wear protective gloves/protective clothing/eye protection/face protection.

IF exposed or concerned: Get medical advice/attention.

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only