Learn More
Thermo Scientific Chemicals Raloxifene hydrochloride
CAS: 82640-04-8 | C28H28ClNO4S, C28H27NO4S·HCl | 510.05 g/mol
Supplier: Thermo Scientific Chemicals 466500050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Raloxifene hydrochloride | |
| 5 g | |
| 263.0°C | |
| 510.04 | |
| [H+].[Cl-].OC1=CC=C(C=C1)C1=C(C(=O)C2=CC=C(OCCN3CCCCC3)C=C2)C2=CC=C(O)C=C2S1 | |
| 510.05 |
| 82640-04-8 | |
| Solubility aq. soln.: .. Other solubilities: soluble in dmso | |
| C28H27NO4S·HCl | |
| BKXVVCILCIUCLG-UHFFFAOYSA-N | |
| hydrogen 2-(4-hydroxyphenyl)-3-{4-[2-(piperidin-1-yl)ethoxy]benzoyl}-1-benzothiophen-6-ol chloride |
Chemical Identifiers
| 82640-04-8 | |
| C28H27NO4S·HCl |
Safety and Handling
GHS H Statement Suspected of causing cancer.
May damage fertility or the unborn child.
GHS P Statement Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
GHS Signal Word: Danger
RUO – Research Use Only