accessibility menu, dialog, popup

UserName

CAS RN 82640-04-8

HClHOSOHOON

CAS RN 82640-04-8

IUPAC Name: hydrogen 2-(4-hydroxyphenyl)-3-{4-[2-(piperidin-1-yl)ethoxy]benzoyl}-1-benzothiophen-6-ol chloride
Synonyms: evista raloxifene hydrochloride
Molecular Weight (g/mol): 510.05
Molecular Formula: C28H28ClNO4S
InChi Key: BKXVVCILCIUCLG-UHFFFAOYSA-N
SMILES: [H+].[Cl-].OC1=CC=C(C=C1)C1=C(C(=O)C2=CC=C(OCCN3CCCCC3)C=C2)C2=CC=C(O)C=C2S1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results