Learn More
Thermo Scientific™ Specialty Racks
Maximize the storage space in your ultra-low temperature freezer with these racks.
$519.38 - $951.00
| Catalog Number | Mfr. No. | Holds | Positions | Product Type | For Use With (Equipment) | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Holds | Positions | Product Type | For Use With (Equipment) | Price | Quantity & Availability | ||
|
1950652
|
Thermo Scientific™
1950652 |
Microplates | 84 Plates/Rack | Side Access Freezer Rack (Microplate/Deepwell) | 17.3, 23 cu. ft. Upright Freezers |
Each for $775.74
|
|
||
|
820036
|
Thermo Scientific™
820036 |
Microplates | 36 Plates/Rack | Side Access Freezer Rack (Microplate/Deepwell) | 17.3, 23 cu. ft. Upright Freezers |
Each for $864.68
|
|
||
|
920015
|
Thermo Scientific™
920015 |
Boxes (2 in.) | 15 Boxes/Rack | Side Access Freezer Rack (Adjustable) | 13 cu. ft. Upright Freezers |
Each for $564.74
|
|
||
|
920017
|
Thermo Scientific™
920017 |
Boxes (3 in.) | 12 Boxes/Rack | Side Access Freezer Rack (Adjustable) | 17.3, 23 cu. ft. Upright Freezers |
Each for $651.62
|
|
||
|
920018
|
Thermo Scientific™
920018 |
Boxes (2 in.) | 20 Boxes/Rack | Side Access Freezer Rack (Adjustable) | 17.3, 23 cu. ft. Upright Freezers |
Each for $651.62
|
|
||
|
920022
|
Thermo Scientific™
920022 |
Boxes (2 in.) | 22 Boxes/Rack | Side Access Freezer Rack (Adjustable) | 17.3, 23 cu. ft. Upright Freezers |
Each for $651.62
|
|
||
|
920094
|
Thermo Scientific™
920094 |
Boxes (2 in.) | Box Racks - Sliding Drawer | 13 cu. ft. Upright Freezers |
Each for $519.38
|
|
|||
|
920095
|
Thermo Scientific™
920095 |
Boxes (3 in.) | 9 Boxes/Rack | Box Racks - Sliding Drawer | 13 cu. ft. Upright Freezers |
Each for $556.47
|
|
||
|
920096
|
Thermo Scientific™
920096 |
Boxes (2 in.) | Box Racks - Sliding Drawer | 17.3, 23 cu. ft. Upright Freezers |
Each for $646.17
|
|
|||
|
920097
|
Thermo Scientific™
920097 |
Boxes (3 in.) | 12 Boxes/Rack | Box Racks - Sliding Drawer | 17.3, 23 cu. ft. Upright Freezers |
Each for $662.98
|
|
||
|
13994163
|
Thermo Scientific™
398329 |
Boxes (2 in.) | 20 Boxes/Rack | Box Racks - Sliding Drawer | Thermo Scientific TDE and TSX Series |
Each for $951.00
|
|
||
|
1950518
|
Thermo Scientific™
1950518 |
Boxes (2 in.) | 20 Boxes/Rack | Side Access Freezer Rack (Adjustable) | Thermo Scientific TDE and TSX Series (5 shelves) |
Each for $682.64
|
|
||
|
1950519
|
Thermo Scientific™
1950519 |
Boxes (3 in.) | 15 Boxes/Rack | Side Access Freezer Rack (Adjustable) | Thermo Scientific TDE and TSX Series (5 shelves) |
Each for $890.17
|
|
||
|
13994156
|
Thermo Scientific™
398322 |
Microplates or Deepwell Microplates | 98 microplates with lids, 119 microplates without lids, and 35 deepwell | Side Access Freezer Rack (Microplate/Locking Rod) | 32 cu.ft. Revco Upright ULT |
Each for $628.87
|
|
||
|
398328
|
Thermo Scientific™
398328 |
Boxes (3 in.) | 15 Boxes/Rack | Box Racks - Sliding Drawer | Thermo Scientific TDE and TSX Series (5 shelves) |
Each for $688.28
|
|
||
Description
Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for previous generation Forma™ 900, 7000 Series, Revco™ UxF, HERAfreeze™ HFU T and TSE/TSD Series Freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.
- With sliding drawer racks
- Adjustable side access box racks
- Microplate racks in standard or deepwell formats
- Warranty varies by country, please contact us for details