Learn More
Thermo Scientific™ Specialty Racks
Maximize the storage space in your ultra-low temperature freezer with these racks.
Supplier:  Thermo Scientific™ 1950519
 View more versions of this product
Catalog No. 1950519
Get Thermo Scientific Accessories with Purchase
View all promotions
Only null left
Description
Description
Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for previous generation Forma™ 900, 7000 Series, Revco™ UxF, HERAfreeze™ HFU T and TSE/TSD Series Freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.
  • Warranty varies by country, please contact us for details
Specifications
Specifications
Silver
Stainless Steel
Thermo Scientific TDE and TSX Series (5 shelves)
9.4 in.
68.1 cm
4 Tiers
13.7 cm
Boxes (3 in.)
15 Boxes/Rack
5.4 in.
26.8 in.
Side Access Freezer Rack (Adjustable)
23.9 cm
Product Suggestions
Product Suggestions