Learn More
Identifiants chimiques
| CAS | 143900-43-0 |
|---|---|
| Molecular Formula | C10H19NO3 |
| Molecular Weight (g/mol) | 201.26 |
| InChI Key | UIJXHKXIOCDSEB-MRVPVSSYSA-N |
| Synonym | r-1-boc-3-hydroxypiperidine, r-tert-butyl 3-hydroxypiperidine-1-carboxylate, tert-butyl 3r-3-hydroxypiperidine-1-carboxylate, r-1-n-boc-3-hydroxypiperidine, r-n-boc-3-hydroxypiperidine, r-1-boc-3-hydroxylpiperidine, 3r-1-tert-butoxycarbonyl-3-hydroxypiperidine, 1-piperidinecarboxylic acid, 3-hydroxy-, 1,1-dimethylethyl ester, 3r, r-3-hydroxy-piperidine-1-carboxylic acid tert-butyl ester, r-3-hydroxy-1-tert-butoxycarbonyl piperidine |
| PubChem CID | 1514398 |
| IUPAC Name | tert-butyl (3R)-3-hydroxypiperidine-1-carboxylate |
| SMILES | CC(C)(C)OC(=O)N1CCCC(C1)O |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantity & Availability | ||||
|
AC436430050
|
N/A | 5g | Glass bottle |
N/A
|
N/A | ||||
| Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identifiants chimiques
| 143900-43-0 | |
| 201.26 | |
| r-1-boc-3-hydroxypiperidine, r-tert-butyl 3-hydroxypiperidine-1-carboxylate, tert-butyl 3r-3-hydroxypiperidine-1-carboxylate, r-1-n-boc-3-hydroxypiperidine, r-n-boc-3-hydroxypiperidine, r-1-boc-3-hydroxylpiperidine, 3r-1-tert-butoxycarbonyl-3-hydroxypiperidine, 1-piperidinecarboxylic acid, 3-hydroxy-, 1,1-dimethylethyl ester, 3r, r-3-hydroxy-piperidine-1-carboxylic acid tert-butyl ester, r-3-hydroxy-1-tert-butoxycarbonyl piperidine | |
| tert-butyl (3R)-3-hydroxypiperidine-1-carboxylate |
| C10H19NO3 | |
| UIJXHKXIOCDSEB-MRVPVSSYSA-N | |
| 1514398 | |
| CC(C)(C)OC(=O)N1CCCC(C1)O |
Spécifications
| 143900-43-0 | |
| 100.0 | |
| Authentic | |
| C10H19NO3 | |
| r-1-boc-3-hydroxypiperidine, r-tert-butyl 3-hydroxypiperidine-1-carboxylate, tert-butyl 3r-3-hydroxypiperidine-1-carboxylate, r-1-n-boc-3-hydroxypiperidine, r-n-boc-3-hydroxypiperidine, r-1-boc-3-hydroxylpiperidine, 3r-1-tert-butoxycarbonyl-3-hydroxypiperidine, 1-piperidinecarboxylic acid, 3-hydroxy-, 1,1-dimethylethyl ester, 3r, r-3-hydroxy-piperidine-1-carboxylic acid tert-butyl ester, r-3-hydroxy-1-tert-butoxycarbonyl piperidine | |
| CC(C)(C)OC(=O)N1CCCC(C1)O | |
| 201.26 | |
| 201.26 | |
| (R)-1-BOC-3-Hydroxypiperidine |
| 96.0 | |
| 104.4°C | |
| 96% min. (GC) | |
| -8.5° to -12.0° (20°C, 589nm) (c=1 in CHCl3) | |
| UIJXHKXIOCDSEB-MRVPVSSYSA-N | |
| tert-butyl (3R)-3-hydroxypiperidine-1-carboxylate | |
| 1514398 | |
| 97% |
Sécurité et manipulation
GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Danger
RUO – Research Use Only