Learn More
(R)-1-BOC-3-hydroxypiperidine, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 436430050
| Quantity | 5g |
|---|---|
| Packaging | Glass bottle |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 143900-43-0 | |
| 201.26 | |
| r-1-boc-3-hydroxypiperidine, r-tert-butyl 3-hydroxypiperidine-1-carboxylate, tert-butyl 3r-3-hydroxypiperidine-1-carboxylate, r-1-n-boc-3-hydroxypiperidine, r-n-boc-3-hydroxypiperidine, r-1-boc-3-hydroxylpiperidine, 3r-1-tert-butoxycarbonyl-3-hydroxypiperidine, 1-piperidinecarboxylic acid, 3-hydroxy-, 1,1-dimethylethyl ester, 3r, r-3-hydroxy-piperidine-1-carboxylic acid tert-butyl ester, r-3-hydroxy-1-tert-butoxycarbonyl piperidine | |
| tert-butyl (3R)-3-hydroxypiperidine-1-carboxylate |
| C10H19NO3 | |
| UIJXHKXIOCDSEB-MRVPVSSYSA-N | |
| 1514398 | |
| CC(C)(C)OC(=O)N1CCCC(C1)O |
Specifications
| (R)-1-BOC-3-Hydroxypiperidine | |
| 96.0 | |
| 104.4°C | |
| 96% min. (GC) | |
| C10H19NO3 | |
| -8.5° to -12.0° (20°C, 589nm) (c=1 in CHCl3) | |
| UIJXHKXIOCDSEB-MRVPVSSYSA-N | |
| tert-butyl (3R)-3-hydroxypiperidine-1-carboxylate | |
| 1514398 | |
| 97% |
| 143900-43-0 | |
| 100.0 | |
| Authentic | |
| Glass bottle | |
| 5g | |
| r-1-boc-3-hydroxypiperidine, r-tert-butyl 3-hydroxypiperidine-1-carboxylate, tert-butyl 3r-3-hydroxypiperidine-1-carboxylate, r-1-n-boc-3-hydroxypiperidine, r-n-boc-3-hydroxypiperidine, r-1-boc-3-hydroxylpiperidine, 3r-1-tert-butoxycarbonyl-3-hydroxypiperidine, 1-piperidinecarboxylic acid, 3-hydroxy-, 1,1-dimethylethyl ester, 3r, r-3-hydroxy-piperidine-1-carboxylic acid tert-butyl ester, r-3-hydroxy-1-tert-butoxycarbonyl piperidine | |
| CC(C)(C)OC(=O)N1CCCC(C1)O | |
| 201.26 | |
| 201.26 |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Danger
Promotions