missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ QuantStudio™ 3D Digital PCR Master Mix
Optimized for use with isolated DNA or cDNA, TaqMan™ assays, and the QuantStudio™ 3D Digital PCR Chip
Specifications
| Concentration | 2X |
|---|---|
| Content And Storage | Remove the tube from the box, then store it at -20°C until first use, then store it at 2° to 8°C. |
| Detection Method | Primer-probe |
| For Use With (Equipment) | v1 chips |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
4482710
|
Applied Biosystems™
4482710 |
1.5mL |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
4485718
|
Applied Biosystems™
4485718 |
5mL |
N/A
|
|
|||||
Specifications
| 2X | |
| Remove the tube from the box, then store it at -20°C until first use, then store it at 2° to 8°C. | |
| Primer-probe | |
| v1 chips |
For Research Use Only. Not for use in diagnostic procedures.