missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ QuantStudio™ 3D Digital PCR Master Mix

Catalog No. 4485718
Encompass
Change view
Click to view available options
Quantity:
1.5mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4485718 5mL
4482710 1.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 4485718 Supplier Applied Biosystems™ Supplier No. 4485718

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Optimized for use with isolated DNA or cDNA, TaqMan™ assays, and the QuantStudio™ 3D Digital PCR Chip

Specifications

Concentration 2X
Content And Storage Remove the tube from the box, then store it at -20°C until first use, then store it at 2° to 8°C.
Detection Method Primer-probe
For Use With (Equipment) v1 chips
Quantity 5mL

For Research Use Only. Not for use in diagnostic procedures.