Learn More
Applied Biosystems™ QuantStudio 3 0.1 mL and TaqMan Master Mix Starter Package
Catalog No. A38706
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Capacité d'absorption:
1 x 96-well Plate
Catalog No. Capacité d'absorption
A38706 1 x 96-well Plate
1 missing translation for 'options'
Catalog No. A38706 missing translation for 'mfr' Applied Biosystems™ missing translation for 'supplierNo' A38706
Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Complete qPCR workflow bundle featuring QuantStudio 3 0.1 mL system with laptop, SuperScript IV VILO Master Mix with ezDNase, TaqMan Fast Advanced Master Mix, 96-well fast plates, optical adhesive covers, and SmartStart setup for fast, precise gene expression analysis.
Enable fast, precise, and reproducible real-time PCR with this comprehensive QuantStudio™ 3 0.1 mL qPCR workflow bundle. Designed to support high-performance gene expression analysis, this package combines advanced instrumentation, optimized reagents, and essential consumables—along with a dedicated laptop and SmartStart onboarding support—to streamline implementation and accelerate results.
This all-in-one solution is ideal for laboratories seeking a complete, compatible workflow for TaqMan™-based assays, reducing the complexity of sourcing individual components while ensuring consistent performance across every step of the qPCR process.
What's Included
  • QuantStudio™ 3 0.1 mL Real-Time PCR System
  • Pre-configured laptop for instrument control and data analysis
  • SmartStart installation and onboarding support for QuantStudio™ 3
  • SuperScript™ IV VILO™ Master Mix with ezDNase™
  • TaqMan™ Fast Advanced Master Mix
  • 96-well fast thermal cycling plates
  • MicroAmp™ optical adhesive covers
Features
  • Complete qPCR workflow solution: Includes instrument, laptop, reagents, and consumables in a single bundle to simplify procurement and ensure compatibility.
  • High-performance real-time PCR system: The QuantStudio™ 3 system delivers precise thermal cycling, sensitive fluorescence detection, and intuitive software for reliable data acquisition and analysis.
  • Fast cycling capability: 0.1 mL block format and fast thermal cycling plates enable rapid run times without compromising data quality.
  • Accelerated onboarding with SmartStart: Guided installation and setup support help users quickly begin generating results with confidence.
  • Optimized reverse transcription: SuperScript™ IV VILO™ Master Mix with ezDNase™ supports efficient cDNA synthesis and helps reduce genomic DNA contamination.
  • High-specificity probe-based chemistry: TaqMan™ Fast Advanced Master Mix enables sensitive, precise quantification with reduced background and high reproducibility.
  • Efficient plate-based workflow: 96-well fast plates support streamlined sample processing for routine and higher-throughput applications.
  • Reliable sealing for consistent results: Optical adhesive covers minimize evaporation and ensure uniform fluorescence detection across wells.
Benefits
  • Accelerate time to data with a fully integrated system designed for fast setup and rapid thermal cycling.
  • Simplify purchasing and setup by consolidating all essential qPCR components into one convenient SKU.
  • Enhance data accuracy and specificity using TaqMan™ probe-based chemistry for precise gene expression analysis.
  • Improve reproducibility with optimized reagents and instrumentation designed to work seamlessly together.
  • Increase laboratory efficiency by reducing workflow complexity and minimizing preparation steps.
  • Support a range of applications including gene expression analysis, assay validation, and routine real-time PCR experiments.
This QuantStudio™ 3 0.1 mL qPCR workflow bundle with TaqMan™ chemistry provides a reliable, high-performance solution for laboratories looking to streamline operations and generate consistent, high-quality real-time PCR results.
Specifications
Format de bloc Non-interchangeable
Capacité d'absorption 1 x 96-well Plate
Tension 100/240 V
Type de produit Real Time PCR Instrument
Nb de programmes 1000
Configurations des blocs 96-well FAST