Learn More
Applied Biosystems™ QuantStudio 3 0.1 mL and SYBR Master Mix Starter Package
Description
Streamline your real-time PCR workflow with this comprehensive, ready-to-run solution designed for reliable, high-performance gene expression analysis. The QuantStudio™ 3 0.1 mL qPCR System package combines advanced instrumentation, optimized reagents, and essential consumables—paired with a dedicated laptop and SmartStart setup—to enable fast implementation and consistent results from day one.
Ideal for research laboratories seeking efficiency, sensitivity, and reproducibility, this bundle eliminates the need to source individual components while ensuring full compatibility across the entire qPCR workflow.
What's Included
- QuantStudio™ 3 0.1 mL Real-Time PCR System
- Pre-configured laptop for instrument control and data analysis
- SmartStart installation and onboarding support for QuantStudio™ 3
- SuperScript™ IV VILO™ Master Mix with ezDNase™
- PowerUp™ SYBR™ Green Master Mix (5 x 5 mL)
- 96-well fast thermal cycling plates
- MicroAmp™ optical adhesive covers
Features
- Complete, integrated workflow: Includes instrument, software-ready laptop, reagents, and consumables in one convenient package.
- High-performance qPCR system: QuantStudio™ 3 delivers precise thermal control, sensitive fluorescence detection, and intuitive software for dependable data collection and analysis.
- Fast implementation: SmartStart support helps accelerate installation, setup, and onboarding so labs can begin running experiments quickly.
- Optimized reverse transcription: SuperScript™ IV VILO™ Master Mix with ezDNase™ supports efficient cDNA synthesis while reducing genomic DNA carryover.
- Robust SYBR Green chemistry: PowerUp™ SYBR™ Green Master Mix supports strong amplification performance, specificity, and reproducibility.
- Efficient plate-based workflow: 96-well fast thermal cycling plates support rapid cycling and streamlined sample processing.
- Reliable sealing for consistent results: Optical adhesive covers help minimize evaporation and support accurate fluorescence detection.
Benefits
- Accelerate lab readiness with a bundled solution that reduces setup time and simplifies implementation.
- Reduce purchasing complexity by sourcing a compatible qPCR workflow under a single SKU.
- Improve confidence in results with trusted instrumentation, optimized chemistries, and workflow-ready consumables.
- Support reproducibility through integrated components designed to work together across the qPCR process.
- Increase productivity with a complete package suitable for gene expression analysis and routine qPCR applications.
This all-in-one qPCR workflow bundle is designed to help laboratories simplify procurement, reduce implementation time, and generate high-quality real-time PCR results with confidence.
Specifications
Specifications
| Block Format | Non-interchangeable |
| Capacity | 1 x 96-well Plate |
| Voltage | 100/240 V |
| Product Type | Real Time PCR Instrument |
| No. of Programs | 1000 |
| Block Configurations | 96-well FAST |