missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quality Control Standards 21 For ICP, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC QC21250
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| As | 100μg/mL |
| Be | 100μg/mL |
| Ca | 100μg/mL |
| Cd | 100μg/mL |
| Co | 100μg/mL |
| Cr | 100μg/mL |
| Cu | 100μg/mL |
| Fe | 100μg/mL |
| Li | 100μg/mL |
| Mg | 100μg/mL |
| Mn | 100μg/mL |
| Mo | 100μg/mL |
| Ni | 100μg/mL |
| Pb | 100μg/mL |
| Sb | 100μg/mL |
| Se | 100μg/mL |
| Sr | 100μg/mL |
| Ti | 100μg/mL |
| Tl | 100μg/mL |
| V | 100μg/mL |
| Zn | 100μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
| Quality Control Standard 21 | |
| Triple-leached LDPE bottle | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| As,Be,Ca | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| Be, Ca, Ca | |
| 250 mL |
Product intended for laboratory use only