missing translation for 'onlineSavingsMsg'
Learn More

Pyridoxamine dihydrochloride, 98%, MilliporeSigma™ Supelco™

$176.52 - $2436.88

Specifications

CAS 524-36-7
Physical Form Neat
Linear Formula C8H12N2O2 · 2 HCl
MDL Number MFCD00012808
Melting Point 224°C to 226°C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111014451
View Documents
Merck Emd Millipore
P93801G
1 g
Each for $176.52
Only null left
 
111014453
View Documents
Merck Emd Millipore
P93805G
5 g
Each for $604.84
Only null left
 
111014452
View Documents
Merck Emd Millipore
P938025G
25 g
Each for $2,436.88
Only null left
 
Description

Description

Pyridoxamine dihydrochloride is a salt of pyridoxamine, which belongs to the family of vitamin B6 compounds. Pyridoxamine occurs naturally in animal-based food products. Its deficiency in humans potentially causes sideroblastic anemia, weakness, insomnia, and neurological disorders. It has been reported to have been effective against diabetic nephropathy. Featured Industries: Vitamins, Nutraceuticals and Natural Products
Specifications

Specifications

524-36-7
C8H12N2O2 · 2 HCl
224°C to 226°C
Analytical Standard
≥98%
Neat
MFCD00012808
4-(Aminomethyl)-5-hydroxy-6-methyl-3-pyridinemethanol dihydrochloride
241.11
Analytical Standard
Videos
SDS
Documents

Documents