missing translation for 'onlineSavingsMsg'
Learn More

Pyridoxamine dihydrochloride, 98%, MilliporeSigma™ Supelco™

Catalog No. 111014451
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014451 1 g
111014453 5 g
111014452 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111014451 Supplier Merck Emd Millipore Supplier No. P93801G
Pyridoxamine dihydrochloride is a salt of pyridoxamine, which belongs to the family of vitamin B6 compounds. Pyridoxamine occurs naturally in animal-based food products. Its deficiency in humans potentially causes sideroblastic anemia, weakness, insomnia, and neurological disorders. It has been reported to have been effective against diabetic nephropathy. Featured Industries: Vitamins, Nutraceuticals and Natural Products

Specifications

CAS 524-36-7
Physical Form Neat
Linear Formula C8H12N2O2 · 2 HCl
MDL Number MFCD00012808
Melting Point 224°C to 226°C
Synonym 4-(Aminomethyl)-5-hydroxy-6-methyl-3-pyridinemethanol dihydrochloride
Type Analytical Standard
Quantity 1 g
Formula Weight 241.11
Percent Purity ≥98%
Grade Analytical Standard
Show More Show Less