missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridoxamine dihydrochloride, 98%, MilliporeSigma™ Supelco™
Supplier: Merck Emd Millipore P938025G
Description
Pyridoxamine dihydrochloride is a salt of pyridoxamine, which belongs to the family of vitamin B6 compounds. Pyridoxamine occurs naturally in animal-based food products. Its deficiency in humans potentially causes sideroblastic anemia, weakness, insomnia, and neurological disorders. It has been reported to have been effective against diabetic nephropathy. Featured Industries: Vitamins, Nutraceuticals and Natural ProductsSpecifications
| 524-36-7 | |
| C8H12N2O2 · 2 HCl | |
| 224°C to 226°C | |
| Analytical Standard | |
| 241.11 | |
| Analytical Standard |
| Neat | |
| MFCD00012808 | |
| 4-(Aminomethyl)-5-hydroxy-6-methyl-3-pyridinemethanol dihydrochloride | |
| 25 g | |
| ≥98% |