Learn More
Corning™ PYREX™ Reusable Petri Dishes: Covers Only
For use with Corning PYREZ Brand Reusable Petri Dishes
Supplier:  Corning™ 3160102CO
 Afficher plus de versions de ce produit
Numéro de catalogue. 08749D
Il en reste null
Description
Description
These flat, clear PYREX™ dish covers will withstand repeated sterilization (wet or dry). The edges are beaded to provide greater mechanical strength. The bead also provides a means to equally space the side walls of the bottom and cover, thereby reducing the capillary action of the condensed moisture on the sides. They are not affected chemically or thermally by any of the methods commonly employed in laboratories where sterilization is routinely used. The covers are marked in blue enamel.
Spécifications
Identifiants chimiques
Reusable
Round
Cell Culture Petri Dish
12/Cs.
Borosilicate Glass
Sterile
Beaded
12/Pk.
Spécifications
Clear
Borosilicate Glass
Sterile
Beaded
12/Cs.
20 mm
Reusable
Round
Cell Culture Petri Dish
08-747D
12/Pk.
100 mm
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS