missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ PYREX™ Reagent Bottles with Hollow standard taper Stoppers
For storing reagents, sera, vaccines, colloidal solutions and similar liquids because of their outstanding chemical durability
$92.39 - $4,675.40
Specifications
| Material | Borosilicate Glass |
|---|---|
| Sterility | Non-sterile |
| Closure Color | Orange |
| Mouth | Narrow |
| Closure Material | Polypropylene |
Alert:
These bottles should never be used pressurized or under vacuum or heated on hotplates or over Bunsen burners.
| Catalog Number | Mfr. No. | Capacity (Metric) | Stopper Number | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Stopper Number | Price | Quantity & Availability | ||||
|
02940B
|
Corning™
1500125 |
125 mL | 19 |
Pack of 6 Each for $464.61
Case of 8 Pack for $3,716.88
|
|
||||
|
02940C
|
Corning™
1500250 |
250 mL | 19 |
Each for $106.26
Case of 36 Each for $3,825.36
|
|
||||
|
02940D
|
Corning™
1500500 |
500 mL | 24 |
Each for $92.39
Case of 24 Each for $2,217.36
|
|
||||
|
02940E
|
Corning™
15001L |
1 L | 29 |
Case of 24 Each for $4,675.40
|
|
||||
|
02940F
|
Corning™
15002L |
2000 mL | 29 |
Case of 6 Each for $2,382.88
|
|
||||
Description
With neck ground and furnished with standard taper stopper.
For stopper only, see Cat. No. 7575.
Order Info
Replacement stoppers available (No. 7575 series).
Specifications
| Borosilicate Glass | |
| Orange | |
| Polypropylene |
| Non-sterile | |
| Narrow |