missing translation for 'onlineSavingsMsg'
Learn More

Corning™ PYREX™ Reagent Bottles with Hollow standard taper Stoppers

For storing reagents, sera, vaccines, colloidal solutions and similar liquids because of their outstanding chemical durability

Supplier:  Corning™ 1500500

 View more versions of this product

Catalog No. 02940D



Only null left

Alert:

These bottles should never be used pressurized or under vacuum or heated on hotplates or over Bunsen burners.


Description

Description

With neck ground and furnished with standard taper stopper.

For stopper only, see Cat. No. 7575.

Order Info

Replacement stoppers available (No. 7575 series).

Specifications

Chemical Identifiers

Borosilicate Glass
Orange
Polypropylene
Non-sterile
Narrow

Specifications

16.9 oz., 500 mL
16.9 oz.
1.39 in.
Borosilicate Glass
1/Pk.
24
Narrow
198 mm
Polypropylene
Screw Cap
500 mL
3.38 in.
35.5 mm
24/Cs.
Non-sterile
Orange
Cylindrical
7.79 in.
86 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions