missing translation for 'onlineSavingsMsg'
Learn More

Corning™ PYREX™ Reagent Bottles with Hollow standard taper Stoppers

Catalog No. 02940F
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1 L
2000 mL
Stopper Number:
19
24
29
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Stopper Number
02940F 2000 mL 29
02940B 125 mL 19
02940C 250 mL 19
02940D 500 mL 24
02940E 1 L 29
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
These bottles should never be used pressurized or under vacuum or heated on hotplates or over Bunsen burners.
Catalog No. 02940F Supplier Corning™ Supplier No. 15002L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

These bottles should never be used pressurized or under vacuum or heated on hotplates or over Bunsen burners.

For storing reagents, sera, vaccines, colloidal solutions and similar liquids because of their outstanding chemical durability

With neck ground and furnished with standard taper stopper.

For stopper only, see Cat. No. 7575.

Order Info

Replacement stoppers available (No. 7575 series).

Specifications

Capacity 67.62 oz., 2000 mL
Capacity (Metric) 2000 mL
Diameter (English) 5.23 in.
Diameter (English) Outer Neck 1.77 in.
Diameter (Metric) Outer Neck 45 mm
Material Borosilicate Glass
No. per Case 6/Cs.
No. per Pack 1/Pk.
Sterility Non-sterile
Stopper Number 29
Closure Color Orange
Mouth Narrow
Height (Metric) 282 mm
Height (English) 11 in.
Closure Material Polypropylene
Diameter (Metric) 133 mm
Closure Type Screw Cap
Capacity (English) 67.62 oz.
Shape Cylindrical
Show More Show Less