missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ PYREX™ Reagent Bottles with Hollow standard taper Stoppers
For storing reagents, sera, vaccines, colloidal solutions and similar liquids because of their outstanding chemical durability
Supplier: Corning™ 15001L
Alert:
These bottles should never be used pressurized or under vacuum or heated on hotplates or over Bunsen burners.
Description
With neck ground and furnished with standard taper stopper.
For stopper only, see Cat. No. 7575.
Order Info
Replacement stoppers available (No. 7575 series).
Chemical Identifiers
| Borosilicate Glass | |
| Orange | |
| Polypropylene |
| Non-sterile | |
| Narrow |
Specifications
| 33.81 oz., 1000 mL | |
| 33.81 oz. | |
| 1.77 in. | |
| Borosilicate Glass | |
| 1/Pk. | |
| 29 | |
| Narrow | |
| 235 mm | |
| Polypropylene | |
| Screw Cap |
| 1 L | |
| 4.21 in. | |
| 45 mm | |
| 24/Cs. | |
| Non-sterile | |
| Orange | |
| Cylindrical | |
| 9.25 in. | |
| 107 mm |