missing translation for 'onlineSavingsMsg'
Learn More

Pyrazinamide, 98%

Catalog No. AAJ6682222
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6682222 100 g
AAJ6682209 10 g
AAJ6682214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6682222 Supplier Thermo Scientific Chemicals Supplier No. J6682222

CAS: 98-96-4 | C5H5N3O | 123.115 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 98-96-4
Molecular Formula C5H5N3O
Molecular Weight (g/mol) 123.115
MDL Number MFCD00006132
InChI Key IPEHBUMCGVEMRF-UHFFFAOYSA-N
PubChem CID 1046
ChEBI CHEBI:45285
IUPAC Name pyrazine-2-carboxamide
SMILES C1=CN=C(C=N1)C(=O)N

Specifications

Type Pyrazinamide
Physical Form Powder
Melting Point 189°C to 191°C
Packaging Plastic Bottle
Quantity 100 g
Flash Point >110°C (230°F)
Beilstein 112306
Synonym Pyrazinoic acid amide,Pyrazinecarboxamide
Solubility Information Soluble in water /n
Formula Weight 123.11
Percent Purity 98%
Chemical Name or Material Pyrazinamide
Show More Show Less
EINECSNumber 202-717-6
RTECSNumber UQ2275000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only