missing translation for 'onlineSavingsMsg'
Learn More

Puromycin dihydrochloride, 98%

Catalog No. AAJ61278ME
Change view
Click to view available options
Quantity:
25 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61278ME 500 mg
AAJ61278MB 25 mg
AAJ61278MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ61278ME Supplier Thermo Scientific Chemicals Supplier No. J61278ME

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Puromycin dihydrochloride hydrate, CAS # 58-58-2, is an aminoglycoside showing antibiotic, anti-neoplastic, and anti-protozoal activities. This compound also has the ability to inhibit protein synthesis. | CAS: 58-58-2 | C22H31Cl2N7O5 | 544.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

General Information

• Puromycin dihydrochloride hydrate is the hydrochloride salt form of puromycin and is an aminoglycoside with antibiotic activity isolated from the Streptomyces alboniger
• Puromycin dihydrochloride hydrate is a protein synthesis inhibitor that causes premature chain termination by acting as an analog of the 3’-terminal ends of aminoacyl-tRNA, inhibiting protein synthesis

Applications

• This compound shows anti-microbial, anti-protozoal, anti-trypanosomal, and anti-neoplastic activities and is also used as an antibiotic in cell culture
• It also reveals anti-tumor activity in studies on brain cancer cells
• This compound has been used to study transcriptional regulatory mechanisms that control the sequential and coordinate expression of genes during cell differentiation
• It is active against gram-positive microorganisms, less active against acid-fast bacilli and gram-negative microorganisms. Puromycin can prevent growth of bacteria, protozoa, algae, and mammalian cells in in vitro studies.

Chemical Identifiers

CAS 58-58-2
Molecular Formula C22H31Cl2N7O5
Molecular Weight (g/mol) 544.43
MDL Number MFCD00150080
InChI Key MKSVFGKWZLUTTO-MLYJQVMKNA-N
PubChem CID 131632508
SMILES Cl.Cl.COC1=CC=C(C[C@H](N)C(=O)N[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O)N2C=NC3=C(N=CN=C23)N(C)C)C=C1

Specifications

Type Puromycin Dihydrochloride
Physical Form Powder
Melting Point 168°C to 170°C
Color White
Quantity 500 mg
Beilstein 3853613
Merck Index 14,7943
Solubility Information Soluble in water
IUPAC Name (2S)-2-amino-N-[(2S,3S,4R,5R)-5-[6-(dimethylamino)-9H-purin-9-yl]-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]-3-(4-methoxyphenyl)propanamide dihydrochloride
Formula Weight 544.44
Percent Purity 99%
Chemical Name or Material Puromycin dihydrochloride
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 200-387-8
RTECSNumber AU355000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only