Learn More
Puritan™ HydraFlock™ Flocked Swabs
For collection and release of biological specimens
 Afficher plus de versions de ce produit
Numéro de catalogue. 22025198
Il en reste null
Description
Description
A new era in flock swab technology. HydraFlock provides superior collection and release of specimens. The unique microstructure of Hydra’s multi-length fibers are designed to enhance rapid absorption and quick release of biological specimens.; For buccal cell collection; Specimen collection; Tip material is comprised of polyester bicomponent fiber; Made in USA
Spécifications
Identifiants chimiques
CE, ISO 9001:2008, ISO 13485:2003, FDA GMP's
Sterile
Flocked
Swab
Polystyrene
Spécifications
CE, ISO 9001:2008, ISO 13485:2003, FDA GMP's
6 in.
Swab
15.2 cm
15.2 cm
1.8 cm
0.135 in.
Polystyrene
Flocked
0.098 in.
For Nasopharyngeal and Urethral cell collection, rapid diagnostic testing, specimen collection, forensic evidence collection
0.248 cm
6 in.
0.72 in.
0.34 cm
Sterile
Minitip
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS