Learn More
United Scientific™ Pulley
Ideal for use in demonstrating simple machines.
Supplier:  United Scientific™ PULQ04
 Afficher plus de versions de ce produit
Numéro de catalogue. S15794
Il en reste null
Description
Description
  • Free-running single pulley with 50 mm diameter plastic sheave.
  • Designed with deep V-groove, aluminum frame, and swivel hooks on both ends.
  • Available in single, double, triple, and quadruple pulleys.
Spécifications
Identifiants chimiques
50 mm
Spécifications
Pulley
50 mm
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS