Learn More
United Scientific™ Pulley
Ideal for use in demonstrating simple machines.
| Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
|
S15952
|
United Scientific™
PULS01 |
Single Pulley |
N/A
|
|
|||||
|
S15860
|
United Scientific™
PULD02 |
Double Parallel Pulley |
N/A
|
|
|||||
|
S15840
|
United Scientific™
PULT03 |
Triple Parallel Pulley |
N/A
|
|
|||||
|
S15794
|
United Scientific™
PULQ04 |
Pulley |
N/A
|
|
|||||
Description
- Free-running single pulley with 50 mm diameter plastic sheave.
- Designed with deep V-groove, aluminum frame, and swivel hooks on both ends.
- Available in single, double, triple, and quadruple pulleys.
Specifications
| 50 mm |