Learn More
Promega rNTPs
High-quality rNTPs. Provided at concentrations of 10mM or 100mM in water at neutral pH.
Supplier:  Promega E6000
Encompass
 Afficher plus de versions de ce produit
Numéro de catalogue. PRE6000
Il en reste null
Description
Description
  • High-Quality Ribonucleoside Triphosphates (rNTPs)
  • 400μL rNTP supplied at 10mM or 100mM in water at neutral pH
  • Qualified for use with the Riboprobe(R) and HeLaScribe(R) Systems
  • Purity verified by HPLC
Spécifications
Identifiants chimiques
-30°C to -10°C
100 mM
Provides a cell-free system for in-vitro transcription of eukaryotic genes
Spécifications
rNTP Set (rCTP, rATP, rUTP, rGTP)
100 mM
Provides a cell-free system for in-vitro transcription of eukaryotic genes
-30°C to -10°C
4 x 400 μL
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS