missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega Cell Lysis Solution (CLA)
Cell Lysis Solution is a component of the Wizard™ MagneSil™, Wizard™ Plus and Wizard™ SV 96 and SV 9600 Plasmid DNA Purification Systems and the PureYield Plasmid Midiprep System.
$143.69 - $379.69
Specifications
| Content And Storage | 15°C to 30°C |
|---|---|
| Format | Multiwell |
| For Use With (Equipment) | Wizard™ Magnesil™ plasmid purification system |
| For Use With (Application) | Rapid isolation of plasmid DNA in a 96-well, high-throughput format |
| Quantity | 500 mL |
| Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
PRA7114
|
Promega
A7114 |
Cell Resuspension Solution |
Each for $193.17
|
|
|||||
PRA7132
|
Promega
A7132 |
Neutralization Solution |
Each for $143.69
|
|
|||||
PRA7124
|
Promega
A7124 |
Cell Lysis Solution |
Each for $151.31
|
|
|||||
PRA1655
|
Promega
A1655 |
Elution Buffer |
Each for $379.69
|
|
|||||
Description
- Reagent Used in Plasmid DNA Isolation Protocols
- Cat.# A7122 is a component of the Wizard(R) Plus DNA Purification Systems
- Cat.# A7125 is a component of the PureYield™ Plasmid Midiprep System
- Cat.# A7124 is a component of the Wizard(R) MagneSil(R) Plasmid Purification System
Specifications
| 15°C to 30°C | |
| Wizard™ Magnesil™ plasmid purification system | |
| 500 mL |
| Multiwell | |
| Rapid isolation of plasmid DNA in a 96-well, high-throughput format |