missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega Cell Lysis Solution (CLA)
Cell Lysis Solution is a component of the Wizard™ MagneSil™, Wizard™ Plus and Wizard™ SV 96 and SV 9600 Plasmid DNA Purification Systems and the PureYield Plasmid Midiprep System.
Supplier: Promega A7132
Description
- Reagents Used in DNA Plasmid DNA Purification Protocols
- Cat.# A7131 is a component of the Wizard MagneSil Tfx™ and Wizard(R) Plus DNA Purification Systems
- Cat.# A7132 is a component of the Wizard(R) MagneSil(R) Plasmid Purification System
- Cat.# A1485 is a component of the PureYield™ Plasmid Midiprep System
- Cat.# A1488 is a component of the Wizard(R) SV 96 Plasmid DNA Purification System
Identifiants chimiques
| 15°C to 30°C | |
| Wizard™ Magnesil™ plasmid purification system | |
| 500 mL |
| Multiwell | |
| Rapid isolation of plasmid DNA in a 96-well, high-throughput format |
Spécifications
| 15°C to 30°C | |
| Wizard™ Magnesil™ plasmid purification system | |
| Neutralization Solution |
| Multiwell | |
| Rapid isolation of plasmid DNA in a 96-well, high-throughput format | |
| 500 mL |