missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega Cell Lysis Solution (CLA)
Cell Lysis Solution is a component of the Wizard™ MagneSil™, Wizard™ Plus and Wizard™ SV 96 and SV 9600 Plasmid DNA Purification Systems and the PureYield Plasmid Midiprep System.
Supplier: Promega A1655
Description
- Rapid Isolation of Plasmid DNA in 96-Well Format
- Process multiple plates without user intervention
- Validated automated methods available
Identifiants chimiques
| 15°C to 30°C | |
| Wizard™ Magnesil™ plasmid purification system | |
| 500 mL |
| Multiwell | |
| Rapid isolation of plasmid DNA in a 96-well, high-throughput format |
Spécifications
| 15°C to 30°C | |
| Multiwell | |
| Rapid isolation of plasmid DNA in a 96-well, high-throughput format | |
| 500 mL |
| Elution Buffer | |
| Wizard™ Magnesil™ plasmid purification system | |
| Elution Buffer |